siRNA Details
Gene Symbol: TUBA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207312.2 | NP_997195.1 | 4 | 808 | ||
GenBank mRNA | AA397983.1 | - | 1 | 86 | ||
AK131043.1 | - | 4 | 786 | |||
BC057811.1 | - | 4 | 808 | 5298532 |
Gene Symbol: TUBA3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006001.2 | NP_005992.1 | 4 | 812 | ||
GenBank mRNA | BC011721.2 | - | 4 | 755 | 4025942 |
Gene Symbol: TUBA3FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003608.1 | 3 | 1039 | |||
GenBank mRNA | AK057257.1 | - | 3 | 991 | ||
AK315686.1 | - | 3 | 987 | |||
BC009991.2 | - | 3 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
