siRNA Details
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017802.3 | NP_060272.3 | 12, 13 | 2449 | ||
NR_075098.1 | 12, 13 | 2387 | ||||
GenBank mRNA | AK000404.1 | - | 5, 6 | 946 | ||
AK054595.1 | - | 5, 6 | 1377 | |||
AK056233.1 | - | 12, 13 | 1834 | |||
AK096700.1 | - | 11, 12 | 1796 | |||
AK098430.1 | - | 6, 7 | 969 | |||
AK225507.1 | - | 5, 6 | 946 | |||
AL832914.1 | - | 3, 4 | 446 | |||
BC047240.1 | - | 12, 13 | 2449 | 5274510 | ||
BC094840.1 | - | 5, 6 | 1205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
