siRNA Details
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017802.3 | NP_060272.3 | 13 | 2823 | ||
NR_075098.1 | 13 | 2761 | ||||
GenBank mRNA | AK000404.1 | - | 6 | 1320 | ||
AK054595.1 | - | 6 | 1751 | |||
AK056233.1 | - | 13 | 2208 | |||
AK096700.1 | - | 12 | 2170 | |||
AK098430.1 | - | 7 | 1343 | |||
AK225507.1 | - | 6 | 1320 | |||
AL832914.1 | - | 4 | 820 | |||
BC047240.1 | - | 13 | 2823 | 5274510 | ||
BC094840.1 | - | 6 | 1579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
