siRNA Details
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 3 | 380 | ||
XM_017023136.1 | XP_016878625.1 | 3 | 379 | |||
XM_017023137.1 | XP_016878626.1 | 3 | 322 | |||
GenBank mRNA | AK311978.1 | - | 3 | 289 | ||
AY533031.1 | - | 1 | 139 | |||
BC000790.1 | - | 1 | 304 | 3451361 | ||
BC009065.1 | - | 1 | 339 | |||
BC020922.1 | - | 1 | 343 | 4247157 | ||
BF798008.1 | - | 3 | 251 | |||
BG708388.1 | - | 3 | 413 | |||
BT009827.1 | - | 3 | 283 | |||
CN283809.1 | - | 3 | 413 | |||
CR457072.1 | - | 3 | 283 | |||
D00723.1 | - | 1 | 375 | |||
M69175.1 | - | 3 | 307 |
Gene Symbol: GCSHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033248.1 | 1 | 224 | |||
GenBank mRNA | AK092810.1 | - | 2 | 2108 | ||
AV703937.1 | - | 1 | 353 |
Gene Symbol: LOC729080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033244.1 | 1 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
