siRNA Details
Gene Symbol: EIF4H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022170.1 | NP_071496.1 | 2 | 155 | ||
NM_031992.1 | NP_114381.1 | 2 | 155 | |||
XM_011516556.1 | XP_011514858.1 | 2 | 188 | |||
XM_011516557.1 | XP_011514859.1 | 2 | 173 | |||
GenBank mRNA | AK290676.1 | - | 2 | 175 | ||
AK297651.1 | - | 2 | 200 | |||
AK314749.1 | - | 2 | 257 | |||
BC010021.1 | - | 1 | 153 | 2960402 | ||
BC021214.2 | - | 2 | 153 | 4139578 | ||
BC066928.1 | - | 1 | 172 | 5263716 | ||
D26068.1 | - | 2 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
