siRNA Details
Gene Symbol: DEFB103B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018661.4 | NP_061131.1 | 1 | 238 | ||
GenBank mRNA | AB037972.1 | - | 1 | 43 | ||
AF217245.1 | - | 1 | 20 | |||
AF295370.1 | - | 1 | 55 | |||
AF301470.1 | - | 1 | 238 | |||
AF516673.1 | - | 1 | 20 | |||
AJ237673.1 | - | 1 | 52 |
Gene Symbol: DEFB103A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081551.3 | NP_001075020.1 | 1 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
