siRNA Details
Gene Symbol: FAM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145268.1 | NP_001138740.1 | 1 | 451 | ||
NM_001145269.1 | NP_001138741.1 | 1 | 451 | |||
NR_026879.1 | 1 | 451 | ||||
XM_006715896.3 | XP_006715959.1 | 1 | 457 | |||
XM_011515922.2 | XP_011514224.1 | 1 | 458 | |||
XM_011515923.2 | XP_011514225.1 | 1 | 440 | |||
XM_011515924.2 | XP_011514226.1 | 1 | 458 | |||
XM_011515925.2 | XP_011514227.1 | 1 | 458 | |||
XM_011515926.2 | XP_011514228.1 | 1 | 458 | |||
XM_017011846.1 | XP_016867335.1 | 1 | 458 | |||
XM_017011847.1 | XP_016867336.1 | 1 | 458 | |||
XM_017011848.1 | XP_016867337.1 | 1 | 458 | |||
XM_017011849.1 | XP_016867338.1 | 1 | 440 | |||
GenBank mRNA | AK054745.1 | - | 1 | 361 | ||
AK298753.1 | - | 1 | 375 | |||
BC029175.1 | - | 1 | 432 | |||
DB192367.1 | - | 1 | 451 | |||
DR006842.1 | - | 1 | 371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
