siRNA Details
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 5 | 1542 | ||
NM_182683.2 | NP_872624.1 | 6 | 1377 | |||
NM_182684.2 | NP_872625.1 | 7 | 1457 | |||
GenBank mRNA | AK056760.1 | - | 6 | 1185 | ||
AK290096.1 | - | 5 | 1445 | |||
AY233462.1 | - | 7 | 1290 | |||
BC004304.1 | - | 5 | 1366 | 3625247 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 6 | 592 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 6 | 637 | |||
NR_027776.2 | 5 | 534 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 2 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
