siRNA Details
Gene Symbol: ISCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030940.3 | NP_112202.2 | 4 | 373 | ||
GenBank mRNA | AF284752.1 | - | 4 | 328 | ||
AK055643.1 | - | 4 | 336 | |||
AK225937.1 | - | 4 | 336 | |||
AK296093.1 | - | 4 | 559 | |||
AL834356.1 | - | 4 | 318 | |||
AY015642.1 | - | 4 | 308 | |||
BC002675.2 | - | 4 | 327 | 3608301 | ||
BC053900.1 | - | 4 | 294 | |||
BC071621.1 | - | 4 | 307 | 5107151 | ||
BG772866.1 | - | 4 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
