siRNA Details
Gene Symbol: KRTAP3-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC069448.1 | - | 1 | 97 | 7262684 | |
BC093845.1 | - | 1 | 150 | 7939690 | ||
BC093847.1 | - | 1 | 150 | 7939692 |
Gene Symbol: KRTAP3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031959.2 | NP_114165.1 | 1 | 94 | ||
GenBank mRNA | AJ406932.1 | - | 1 | 94 | ||
BC034582.1 | - | 1 | 102 | 3909612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
