siRNA Details
Gene Symbol: RBAK-RBAKDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204513.1 | NP_001191442.1 | 6 | 756 |
Gene Symbol: RBAKDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015343.2 | 3 | 477 | |||
GenBank mRNA | BC031661.1 | - | 3 | 412 | ||
CR745855.1 | - | 3 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
