siRNA Details
Gene Symbol: FRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005479.3 | NP_005470.2 | 1 | 211 | ||
GenBank mRNA | AB074890.1 | - | 1 | 135 | ||
AK307245.1 | - | 1 | 209 | |||
BC034476.1 | - | 1 | 190 | 4823463 | ||
U58975.2 | - | 1 | 211 |
Gene Symbol: FRAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012083.2 | NP_036215.1 | 1 | 151 | ||
GenBank mRNA | AB045118.1 | - | 1 | 151 | ||
BC020165.1 | - | 1 | 126 | 3629702 | ||
BT009770.1 | - | 1 | 22 | |||
DR006612.1 | - | 1 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
