siRNA Details
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 517 | ||
GenBank mRNA | AY255838.1 | - | 1 | 383 | ||
BC074936.2 | - | 1 | 493 | 30915269 | ||
BC074937.2 | - | 1 | 493 | 30915492 | ||
BC104163.1 | - | 1 | 494 | 40029595 | ||
BC104164.1 | - | 1 | 494 | 40029596 | ||
CR541921.1 | - | 1 | 449 | |||
JN591568.1 | - | 1 | 449 | |||
JN591569.1 | - | 1 | 449 | |||
JN591570.1 | - | 1 | 449 | |||
JN848522.1 | - | 1 | 449 | |||
M54886.1 | - | 1 | 509 | |||
V00544.1 | - | 1 | 509 | |||
V00548.1 | - | 1 | 431 | |||
V00549.1 | - | 1 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
