siRNA Details
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006937.3 | NP_008868.3 | 3 | 372 | ||
GenBank mRNA | AK024823.1 | - | 3 | 273 | ||
AK225090.1 | - | 3 | 283 | |||
AK225263.1 | - | 3 | 284 | |||
AK311837.1 | - | 3 | 318 | |||
BC008450.1 | - | 3 | 339 | 4133768 | ||
BC016775.1 | - | 3 | 353 | 4096440 | ||
BC022340.1 | - | 3 | 323 | 4134531 | ||
BC062713.1 | - | 3 | 286 | 30404344 | ||
BC068465.1 | - | 3 | 338 | 5277638 | ||
BC070159.1 | - | 3 | 346 | 6592382 | ||
BC071645.1 | - | 3 | 279 | 3884132 | ||
BC071646.1 | - | 3 | 315 | 4369354 | ||
BC107853.1 | - | 3 | 276 | 5772748 | ||
BG778261.1 | - | 3 | 373 | |||
L76416.1 | - | 3 | 217 | |||
X99585.1 | - | 3 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
