siRNA Details
Gene Symbol: IQSEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170738.1 | NP_001164209.1 | 1 | 481 | ||
XM_011520958.2 | XP_011519260.1 | 1 | 569 | |||
XM_011520959.2 | XP_011519261.1 | 1 | 569 | |||
XM_011520960.1 | XP_011519262.1 | 1 | 568 | |||
XM_017019310.1 | XP_016874799.1 | 1 | 569 | |||
XM_017019311.1 | XP_016874800.1 | 1 | 569 | |||
GenBank mRNA | BC024764.1 | - | 1 | 599 | ||
BC042067.1 | - | 1 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
