siRNA Details
Gene Symbol: EIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143760.1 | NP_001137232.1 | 3 | 334 | ||
NM_001143761.1 | NP_001137233.1 | 3 | 362 | |||
NM_001143762.1 | NP_001137234.1 | 3 | 418 | |||
NM_001970.4 | NP_001961.1 | 3 | 433 | |||
XM_005256509.2 | XP_005256566.1 | 3 | 353 | |||
GenBank mRNA | AF087992.1 | - | 2 | 147 | ||
AK222585.1 | - | 3 | 232 | |||
AK292615.1 | - | 3 | 270 | |||
AY129319.1 | - | 3 | 334 | |||
AY129320.1 | - | 3 | 421 | |||
AY129321.1 | - | 3 | 346 | |||
AY129322.1 | - | 3 | 390 | |||
BC000751.2 | - | 3 | 320 | 2961296 | ||
BC001832.2 | - | 3 | 320 | 2961296 | ||
BC030160.2 | - | 3 | 308 | 5022012 | ||
BC080196.1 | - | 3 | 303 | 6528111 | ||
BC085015.1 | - | 2 | 228 | 6575132 | ||
BC107779.1 | - | 3 | 312 | 6495558 | ||
BG479628.1 | - | 3 | 422 | |||
CB111036.1 | - | 3 | 433 | |||
M23419.1 | - | 2 | 239 |
Gene Symbol: EIF5AL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099692.1 | NP_001093162.1 | 1 | 245 | ||
GenBank mRNA | BC070048.1 | - | 1 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
