siRNA Details
Gene Symbol: OR7E62P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF359418.1 | - | LIT2087 PJCG8 pseudogene |
1 | 167 | |
AF359419.1 | - | LIT2088 PJCG9 pseudogene |
1 | 166 |
Gene Symbol: LOC650293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040071.1 | NP_001035160.1 | 1 | 418 |
Gene Symbol: OR7E125P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF359416.1 | - | LIT2085 PJCG6 pseudogene |
1 | 166 |
Gene Symbol: OR7E5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027688.2 | 4 | 973 | |||
GenBank mRNA | AK055955.1 | - | 4 | 973 |
Gene Symbol: OR7E91P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002185.3 | 3 | 744 | |||
GenBank mRNA | BC014374.1 | - | 3 | 747 | ||
BC030991.1 | - | 3 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
