siRNA Details
Gene Symbol: VAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014232.2 | NP_055047.2 | 2, 3 | 227 | ||
XM_005256775.4 | XP_005256832.1 | 2, 3 | 557 | |||
GenBank mRNA | AJ225044.1 | - | 1, 2 | 131 | ||
AK021522.1 | - | 2, 3 | 209 | |||
AK225777.1 | - | 2, 3 | 227 | |||
AK289555.1 | - | 2, 3 | 200 | |||
BC002737.2 | - | 2, 3 | 196 | 3628842 | ||
BC019608.1 | - | 2, 3 | 209 | 4508990 | ||
BC033870.1 | - | 2, 3 | 215 | 5203115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
