siRNA Details
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 2 | 468 | ||
NR_126398.1 | 2 | 468 | ||||
XM_017013344.1 | XP_016868833.1 | 2 | 694 | |||
XM_017013345.1 | XP_016868834.1 | 1 | 89 | |||
XM_017013346.1 | XP_016868835.1 | 1 | 89 | |||
GenBank mRNA | AK094450.1 | - | 2 | 416 | ||
BC073830.1 | - | 2 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
