siRNA Details
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 2 | 415 | |||
GenBank mRNA | AK074778.1 | - | 3 | 472 | ||
AK307326.1 | - | 3 | 472 | |||
AK309056.1 | - | 3 | 339 | |||
AK311265.1 | - | 2 | 337 | |||
BC054509.1 | - | 2 | 232 | |||
BC071951.1 | - | 2 | 372 | |||
BC096759.1 | - | 2 | 372 | |||
DQ575135.1 | - | 1 | 14 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC007811.2 | - | 3 | 445 |
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 3 | 494 |
Gene Symbol: LOC642423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308544.1 | - | 3 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
