siRNA Details
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 4 | 640 | |||
GenBank mRNA | AK074778.1 | - | 4 | 605 | ||
AK307326.1 | - | 5 | 697 | |||
AK309056.1 | - | 5 | 564 | |||
AK311265.1 | - | 4 | 562 | |||
BC054509.1 | - | 4 | 457 | |||
BC071951.1 | - | 4 | 597 | |||
BC096759.1 | - | 4 | 597 | |||
BG248101.1 | - | 1 | 51 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 5 | 750 | |||
NR_033936.3 | 4 | 661 | ||||
NR_103496.2 | 4 | 611 | ||||
GenBank mRNA | AK026448.1 | - | 4 | 439 | ||
AK309420.1 | - | 4 | 518 | |||
AK309732.1 | - | 4 | 612 | |||
AK310769.1 | - | 4 | 564 | |||
AK311124.1 | - | 4 | 458 | |||
AK311170.1 | - | 4 | 571 | |||
BC007811.2 | - | 5 | 674 | |||
BC065260.1 | - | 5 | 667 | |||
BC072390.1 | - | 4 | 614 |
Gene Symbol: GOLGA2P2Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001555.2 | 9 | 1020 | |||
GenBank mRNA | AF332229.1 | - | 9 | 1019 |
Gene Symbol: GOLGA2P3Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002195.2 | 9 | 1020 |
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 4 | 627 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 3 | 474 | |||
NR_102748.1 | 3 | 511 | ||||
GenBank mRNA | AK291684.1 | - | 4 | 555 | ||
AK307285.1 | - | 3 | 456 | |||
AK309746.1 | - | 3 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
