siRNA Details
Gene Symbol: VCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016378.3 | NP_057462.2 | 3 | 794 | ||
GenBank mRNA | AF159127.1 | - | 2 | 512 | ||
AF167079.1 | - | 3 | 794 |
Gene Symbol: VCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013452.2 | NP_038480.2 | 3 | 946 | ||
XM_011545490.2 | XP_011543792.1 | 3 | 778 | |||
GenBank mRNA | AF159129.1 | - | 2 | 753 | ||
AF167081.2 | - | 3 | 946 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016379.3 | NP_057463.2 | 3 | 974 | ||
XM_006724494.3 | XP_006724557.1 | 3 | 698 | |||
GenBank mRNA | AF159128.1 | - | 2 | 673 | ||
AF167078.1 | - | 3 | 974 | |||
BC039722.1 | - | 2 | 685 |
Gene Symbol: VCX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF167080.2 | - | 4 | 1064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
