siRNA Details
Gene Symbol: OR2T33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004695.1 | NP_001004695.1 | 1 | 40 | ||
GenBank mRNA | BC136994.1 | - | 1 | 62 | 9020986 | |
BC136995.1 | - | 1 | 62 | 9020987 |
Gene Symbol: OR2T12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004692.1 | NP_001004692.1 | 1 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
