siRNA Details
Gene Symbol: DEFB103B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018661.4 | NP_061131.1 | 2 | 460 | ||
GenBank mRNA | AB037972.1 | - | 2 | 265 | ||
AJ237673.1 | - | 2 | 274 |
Gene Symbol: DEFB103A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081551.3 | NP_001075020.1 | 2 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
