siRNA Details
Gene Symbol: DEFB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205266.1 | NP_001192195.1 | 1 | 51 | ||
GenBank mRNA | BE714502.1 | - | 1 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
