siRNA Details
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130440.1 | NP_001123912.1 | 2 | 225 | ||
NM_003133.5 | NP_003124.1 | 2 | 225 | |||
GenBank mRNA | AF070649.1 | - | 2 | 204 | ||
AK289595.1 | - | 2 | 190 | |||
AL050363.1 | - | 1 | 173 | |||
BC008443.1 | - | 2 | 209 | 4102222 | ||
BC015094.1 | - | 1 | 196 | 3930047 | ||
BC021995.1 | - | 2 | 205 | 4101294 | ||
BC022415.1 | - | 1 | 191 | 4243562 | ||
BC064351.1 | - | 2 | 174 | 4452218 | ||
BC066957.1 | - | 2 | 212 | 5263635 | ||
BC067845.1 | - | 2 | 160 | 5287027 | ||
BC106005.1 | - | 1 | 201 | 6026221 | ||
DA983529.1 | - | 2 | 225 | |||
DB194289.1 | - | 2 | 230 | |||
EF488978.1 | - | 2 | 209 | |||
U20998.1 | - | 2 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
