siRNA Details
Gene Symbol: ANKRD30BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027019.1 | 4 | 842 | |||
NR_027020.2 | 3 | 703 | ||||
GenBank mRNA | BC045801.1 | - | 3 | 703 | ||
BC057764.1 | - | 4 | 845 | |||
BC150521.1 | - | 4 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
