siRNA Details
Gene Symbol: C1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190263.1 | NP_001177192.1 | 3 | 171 | ||
NM_001190265.1 | NP_001177194.1 | 3 | 184 | |||
NM_006333.3 | NP_006324.1 | 2 | 107 | |||
NM_173177.2 | NP_775269.1 | 2 | 94 | |||
GenBank mRNA | AK290451.1 | - | 3 | 154 | ||
BC005235.1 | - | 2 | 78 | 3930648 | ||
BC009584.1 | - | 2 | 70 | 4096552 | ||
BC009589.1 | - | 1 | 64 | 4102498 | ||
BC016284.2 | - | 2 | 63 | 3906517 | ||
BG505686.1 | - | 2 | 107 | |||
DA900336.1 | - | 3 | 161 | |||
X95592.1 | - | 3 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
