siRNA Details
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143985.1 | NP_001137457.1 | 2 | 524 | ||
NM_003860.3 | NP_003851.1 | 2 | 581 | |||
XM_017018514.1 | XP_016874003.1 | 2 | 574 | |||
XM_017018515.1 | XP_016874004.1 | 2 | 114 | |||
GenBank mRNA | AF044773.1 | - | 2 | 177 | ||
AF068235.1 | - | 2 | 580 | |||
AF070447.1 | - | 1 | 73 | |||
AK311961.1 | - | 2 | 148 | |||
BC005942.1 | - | 2 | 160 | 4074168 | ||
BC107702.1 | - | 2 | 257 | 6495215 | ||
BG748852.1 | - | 2 | 364 | |||
BU940708.1 | - | 2 | 341 | |||
CR542140.1 | - | 1 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
