siRNA Details
Gene Symbol: OR6B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173351.1 | NP_775486.1 | 1 | 460 | ||
XM_011510671.2 | XP_011508973.1 | 3 | 857 |
Gene Symbol: OR6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005853.1 | NP_001005853.1 | 1 | 460 | ||
GenBank mRNA | BC137561.1 | - | 1 | 520 | 9021561 | |
BC137562.1 | - | 1 | 520 | 9021562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
