siRNA Details
Gene Symbol: LRRC37A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006607.2 | NP_001006608.2 | 7, 8 | 3053 | ||
XM_011524841.2 | XP_011523143.1 | 6, 7 | 3034 | |||
XM_011524842.2 | XP_011523144.1 | 7, 8 | 3106 | |||
XM_011524844.2 | XP_011523146.1 | 5, 6 | 2953 | |||
XM_011524846.2 | XP_011523148.1 | 4, 5 | 2881 | |||
GenBank mRNA | AK126994.1 | - | 7, 8 | 1735 | ||
AY386262.1 | - | 7, 8 | 1474 | |||
BC144427.1 | - | 7, 8 | 716 | |||
BC144428.1 | - | 7, 8 | 716 | |||
JQ790563.1 | - | 7, 8 | 610 | |||
JQ790564.1 | - | 7, 8 | 610 | |||
JQ790568.1 | - | 7, 8 | 610 | |||
JQ790570.1 | - | 7, 8 | 610 | |||
JQ790571.1 | - | 7, 8 | 610 | |||
JQ790576.1 | - | 7, 8 | 610 | |||
JQ790591.1 | - | 4, 5 | 385 | |||
JQ790595.1 | - | 4, 5 | 385 | |||
JQ790598.1 | - | 6, 7 | 538 | |||
JQ790606.1 | - | 7, 8 | 610 | |||
JQ790610.1 | - | 7, 8 | 610 | |||
JQ790617.1 | - | 7, 8 | 610 | |||
JQ790675.1 | - | 4, 5 | 385 | |||
JQ790677.1 | - | 4, 5 | 385 | |||
JQ790700.1 | - | 7, 8 | 610 | |||
JQ790723.1 | - | 4, 5 | 385 | |||
JQ790739.1 | - | 7, 8 | 612 | |||
JQ790740.1 | - | 7, 8 | 612 | |||
JQ790748.1 | - | 7, 8 | 613 | |||
JQ790750.1 | - | 7, 8 | 612 | |||
JQ790764.1 | - | 7, 8 | 612 |
Gene Symbol: LRRC37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014834.4 | NP_055649.4 | 7, 8 | 3053 | ||
XM_005257879.4 | XP_005257936.1 | 7, 8 | 3311 | |||
XM_005257882.4 | XP_005257939.1 | 8, 9 | 3497 | |||
XM_011525539.2 | XP_011523841.1 | 5, 6 | 3158 | |||
XM_017025472.1 | XP_016880961.1 | 9, 10 | 2910 | |||
XM_017025473.1 | XP_016880962.1 | 7, 8 | 557 | |||
GenBank mRNA | BC040501.1 | - | 7, 8 | 3053 | 5295808 | |
CR749232.1 | - | 7, 8 | 3054 | |||
JQ790562.1 | - | 7, 8 | 610 | |||
JQ790566.1 | - | 7, 8 | 610 | |||
JQ790572.1 | - | 7, 8 | 610 | |||
JQ790577.1 | - | 7, 8 | 610 | |||
JQ790578.1 | - | 7, 8 | 610 | |||
JQ790579.1 | - | 7, 8 | 610 | |||
JQ790580.1 | - | 7, 8 | 610 | |||
JQ790581.1 | - | 7, 8 | 610 | |||
JQ790593.1 | - | 4, 5 | 385 | |||
JQ790594.1 | - | 4, 5 | 385 | |||
JQ790596.1 | - | 4, 5 | 385 | |||
JQ790688.1 | - | 7, 8 | 610 | |||
JQ790690.1 | - | 7, 8 | 610 | |||
JQ790691.1 | - | 7, 8 | 610 | |||
JQ790693.1 | - | 7, 8 | 610 | |||
JQ790694.1 | - | 7, 8 | 610 | |||
JQ790699.1 | - | 7, 8 | 610 | |||
JQ790724.1 | - | 4, 5 | 385 | |||
JQ790725.1 | - | 4, 5 | 385 | |||
JQ790726.1 | - | 7, 8 | 612 | |||
JQ790727.1 | - | 7, 8 | 612 | |||
JQ790729.1 | - | 7, 8 | 612 | |||
JQ790730.1 | - | 7, 8 | 612 | |||
JQ790732.1 | - | 4, 5 | 387 | |||
JQ790738.1 | - | 7, 8 | 612 | |||
JQ790743.1 | - | 7, 8 | 612 | |||
JQ790744.1 | - | 7, 8 | 611 | |||
JQ790749.1 | - | 7, 8 | 612 | |||
JQ790752.1 | - | 7, 8 | 612 | |||
JQ790755.1 | - | 7, 8 | 612 | |||
JQ790756.1 | - | 7, 8 | 612 | |||
JQ790758.1 | - | 7, 8 | 612 | |||
JQ790763.1 | - | 7, 8 | 613 |
Gene Symbol: LRRC37A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JQ790698.1 | - | 7, 8 | 610 |
Gene Symbol: LRRC37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321350.1 | NP_001308279.1 | 10, 11 | 2325 | ||
NM_052888.2 | NP_443120.2 | 7, 8 | 2131 | |||
GenBank mRNA | AJ314647.1 | - | 7, 8 | 2117 | ||
AK299772.1 | - | 10, 11 | 2179 | |||
AK302740.1 | - | 7, 8 | 2262 | |||
AY386261.1 | - | 8, 9 | 2169 | |||
BC117373.1 | - | 5, 6 | 2017 | 40125924 | ||
BC143581.1 | - | 5, 6 | 2017 | 9052092 | ||
BM718301.1 | - | 715 | ||||
JQ790561.1 | - | 7, 8 | 602 | |||
JQ790565.1 | - | 7, 8 | 610 | |||
JQ790567.1 | - | 7, 8 | 610 | |||
JQ790569.1 | - | 7, 8 | 610 | |||
JQ790582.1 | - | 7, 8 | 610 | |||
JQ790601.1 | - | 7, 8 | 610 | |||
JQ790602.1 | - | 7, 8 | 610 | |||
JQ790604.1 | - | 7, 8 | 610 | |||
JQ790615.1 | - | 7, 8 | 610 | |||
JQ790616.1 | - | 7, 8 | 610 | |||
JQ790618.1 | - | 7, 8 | 610 | |||
JQ790619.1 | - | 7, 8 | 610 | |||
JQ790620.1 | - | 7, 8 | 610 | |||
JQ790622.1 | - | 7, 8 | 598 | |||
JQ790672.1 | - | 4, 5 | 385 | |||
JQ790689.1 | - | 7, 8 | 610 | |||
JQ790695.1 | - | 7, 8 | 610 | |||
JQ790696.1 | - | 7, 8 | 610 | |||
JQ790697.1 | - | 7, 8 | 610 | |||
JQ790701.1 | - | 7, 8 | 610 | |||
JQ790707.1 | - | 5, 6 | 457 | |||
JQ790746.1 | - | 7, 8 | 612 | |||
JQ790754.1 | - | 7, 8 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
