siRNA Details
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243718.1 | NP_001230647.1 | 1 | 568 | ||
NM_002869.4 | NP_002860.2 | 1 | 568 | |||
NM_198896.1 | NP_942599.1 | 1 | 568 | |||
GenBank mRNA | AF119836.1 | - | 1 | 116 | ||
AF130122.1 | - | 1 | 57 | |||
AF130986.1 | - | 1 | 47 | |||
AF198616.1 | - | 1 | 47 | |||
AF498939.1 | - | 1 | 47 | |||
AF498941.1 | - | 1 | 47 | |||
AK057157.1 | - | 1 | 533 | |||
AK223551.1 | - | 1 | 486 | |||
AK289748.1 | - | 1 | 486 | |||
AK290132.1 | - | 1 | 217 | |||
BC003617.2 | - | 1 | 464 | 3506585 | ||
BC034996.2 | - | 1 | 429 | |||
BC068486.1 | - | 2 | 509 | 30342562 | ||
BC096818.1 | - | 1 | 502 | 5295149 | ||
BI552973.1 | - | 1 | 503 | |||
BQ422263.1 | - | 1 | 50 | |||
BT019698.1 | - | 1 | 47 | |||
CB989138.1 | - | 536 | ||||
CR541984.1 | - | 1 | 47 | |||
DQ437503.1 | - | 1 | 47 | |||
M28212.1 | - | 1 | 117 | |||
X13904.1 | - | 1 | 270 |
Gene Symbol: RAB6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032144.2 | NP_115520.2 | 1 | 501 | ||
GenBank mRNA | AF309646.1 | - | 1 | 94 | ||
AK055504.1 | - | 1 | 124 | |||
AL136727.1 | - | 1 | 502 | |||
BC120999.1 | - | 1 | 339 | 40116526 | ||
CR533469.1 | - | 1 | 47 |
Gene Symbol: WTH3DI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077637.1 | NP_001071105.1 | 1 | 485 | ||
GenBank mRNA | AB232598.1 | - | 1 | 47 | ||
DB074386.1 | - | 2 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
