siRNA Details
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243719.1 | NP_001230648.1 | 6 | 456 | ||
NM_002869.4 | NP_002860.2 | 6 | 1001 | |||
NM_198896.1 | NP_942599.1 | 6 | 1001 | |||
GenBank mRNA | AF119836.1 | - | 6 | 549 | ||
AF130122.1 | - | 6 | 490 | |||
AF130986.1 | - | 6 | 480 | |||
AF198616.1 | - | 6 | 480 | |||
AF498939.1 | - | 6 | 480 | |||
AF498941.1 | - | 6 | 480 | |||
AK057157.1 | - | 6 | 966 | |||
AK223551.1 | - | 6 | 919 | |||
AK289748.1 | - | 6 | 919 | |||
AK290132.1 | - | 6 | 650 | |||
AK301534.1 | - | 6 | 459 | |||
AK309311.1 | - | 4 | 659 | |||
BC003617.2 | - | 6 | 897 | 3506585 | ||
BC068486.1 | - | 7 | 942 | 30342562 | ||
BC096818.1 | - | 6 | 935 | 5295149 | ||
BQ422263.1 | - | 6 | 483 | |||
BT019698.1 | - | 6 | 480 | |||
M28212.1 | - | 6 | 550 | |||
X13904.1 | - | 6 | 703 |
Gene Symbol: RAB6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032144.2 | NP_115520.2 | 1 | 934 |
Gene Symbol: WTH3DI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077637.1 | NP_001071105.1 | 1 | 918 | ||
GenBank mRNA | AB232598.1 | - | 1 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
