siRNA Details
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 4 | 712 | ||
GenBank mRNA | AK075065.1 | - | 4 | 690 | ||
BC010154.2 | - | 4 | 706 | 3623163 | ||
BC019835.1 | - | 4 | 683 | |||
BC065219.1 | - | 4 | 664 | 5724338 | ||
BC084572.1 | - | 4 | 666 | |||
BC090943.1 | - | 4 | 682 | 5404327 | ||
BX537434.1 | - | 5 | 791 |
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003047.1 | XP_016858536.1 | 4 | 428 | ||
XM_017003048.1 | XP_016858537.1 | 4 | 7123 | |||
XM_017003049.1 | XP_016858538.1 | 4 | 440 | |||
XM_017003050.1 | XP_016858539.1 | 4 | 674 | |||
XM_017003051.1 | XP_016858540.1 | 4 | 673 | |||
XM_017003052.1 | XP_016858541.1 | 5 | 574 |
Gene Symbol: LOC100996763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711700.3 | XP_006711763.1 | 4 | 621 | ||
XM_006711703.3 | XP_006711766.1 | 4 | 683 | |||
XM_011510256.2 | XP_011508558.1 | 4 | 1408 |
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 4 | 758 | ||
NM_024408.3 | NP_077719.2 | 4 | 758 | |||
XM_005270901.3 | XP_005270958.1 | 5 | 3072 | |||
XM_011541519.2 | XP_011539821.1 | 4 | 5866 | |||
XM_011541520.2 | XP_011539822.1 | 4 | 2776 | |||
XM_017001372.1 | XP_016856861.1 | 4 | 428 | |||
XM_017001373.1 | XP_016856862.1 | 4 | 407 | |||
GenBank mRNA | AF308601.1 | - | 4 | 473 | ||
AF315356.1 | - | 4 | 473 | |||
AK307843.1 | - | 3 | 600 | |||
BC071562.1 | - | 4 | 707 | 4822955 | ||
BG718124.1 | - | 4 | 718 | |||
GQ231534.1 | - | 4 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
