siRNA Details
Gene Symbol: LOC101929796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542531.2 | XP_011540833.1 | 4 | 973 | ||
XM_017003012.1 | XP_016858501.1 | 4 | 973 | |||
XM_017003013.1 | XP_016858502.1 | 4 | 973 | |||
XM_017003014.1 | XP_016858503.1 | 4 | 973 |
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003047.1 | XP_016858536.1 | 4 | 698 | ||
XM_017003048.1 | XP_016858537.1 | 4 | 7393 | |||
XM_017003049.1 | XP_016858538.1 | 4 | 710 | |||
XM_017003050.1 | XP_016858539.1 | 4 | 944 | |||
XM_017003051.1 | XP_016858540.1 | 4 | 943 | |||
XM_017003052.1 | XP_016858541.1 | 5 | 844 |
Gene Symbol: LOC100996763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711700.3 | XP_006711763.1 | 4 | 891 | ||
XM_006711703.3 | XP_006711766.1 | 4 | 953 | |||
XM_011510256.2 | XP_011508558.1 | 4 | 1678 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509395.2 | XP_011507697.1 | 1 | 207 |
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 4 | 1028 | ||
NM_024408.3 | NP_077719.2 | 4 | 1028 | |||
XM_005270901.3 | XP_005270958.1 | 5 | 3342 | |||
XM_011541519.2 | XP_011539821.1 | 4 | 6136 | |||
XM_011541520.2 | XP_011539822.1 | 4 | 3046 | |||
XM_017001372.1 | XP_016856861.1 | 4 | 698 | |||
XM_017001373.1 | XP_016856862.1 | 4 | 677 | |||
GenBank mRNA | AF308601.1 | - | 4 | 743 | ||
AF315356.1 | - | 4 | 743 | |||
AK307843.1 | - | 3 | 870 | |||
BC071562.1 | - | 4 | 977 | 4822955 | ||
GQ231534.1 | - | 4 | 890 |
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 4 | 982 | ||
GenBank mRNA | AK075065.1 | - | 4 | 960 | ||
BC010154.2 | - | 4 | 976 | 3623163 | ||
BC019835.1 | - | 4 | 953 | |||
BC065219.1 | - | 4 | 934 | 5724338 | ||
BC084572.1 | - | 4 | 936 | |||
BC090943.1 | - | 4 | 952 | 5404327 | ||
BX537434.1 | - | 5 | 1061 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
