siRNA Details
Gene Symbol: HNRNPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194247.2 | NP_919223.1 | 10 | 1231 | ||
XM_005246380.1 | XP_005246437.1 | 10 | 1231 | |||
XM_005246381.4 | XP_005246438.1 | 10 | 1137 | |||
XM_006712365.2 | XP_006712428.1 | 9 | 1090 | |||
XM_017003587.1 | XP_016859076.1 | 10 | 1137 | |||
XM_017003588.1 | XP_016859077.1 | 9 | 1090 | |||
GenBank mRNA | AF517524.1 | - | 3 | 180 | ||
AK293125.1 | - | 10 | 1099 | |||
AK303208.1 | - | 5 | 437 | |||
AK304687.1 | - | 10 | 995 | |||
AK316434.1 | - | 10 | 1099 | |||
AL832246.1 | - | 4 | 396 | |||
AY363225.1 | - | 10 | 1125 | |||
BC064494.1 | - | 8 | 846 | |||
BC080591.1 | - | 6 | 559 | |||
BC112027.1 | - | 10 | 1153 | 8327495 | ||
BX647217.1 | - | 9 | 875 |
Gene Symbol: HNRNPA3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002726.2 | 1 | 1157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
