siRNA Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004.3 | NP_000995.1 | 2, 3 | 232 | ||
GenBank mRNA | AK130959.1 | - | 2, 3 | 191 | ||
AK311954.1 | - | 2, 3 | 200 | |||
BC005354.1 | - | 2, 3 | 203 | 4052568 | ||
BC005920.1 | - | 2, 3 | 201 | 4274135 | ||
BC007573.1 | - | 2, 3 | 177 | 3049240 | ||
BC062314.1 | - | 2, 3 | 181 | 4685028 | ||
CN270352.1 | - | 2, 3 | 229 | |||
CR542212.1 | - | 1, 2 | 127 | |||
CR542248.1 | - | 1, 2 | 127 | |||
D17174.1 | - | 1, 2 | 53 | |||
M17887.1 | - | 2, 3 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
