siRNA Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004.3 | NP_000995.1 | 2 | 181 | ||
GenBank mRNA | AK130959.1 | - | 2 | 140 | ||
AK311954.1 | - | 2 | 149 | |||
BC005354.1 | - | 2 | 152 | 4052568 | ||
BC005920.1 | - | 2 | 150 | 4274135 | ||
BC007573.1 | - | 2 | 126 | 3049240 | ||
BC062314.1 | - | 2 | 130 | 4685028 | ||
CN270352.1 | - | 2 | 178 | |||
CR542212.1 | - | 1 | 76 | |||
CR542248.1 | - | 1 | 76 | |||
M17887.1 | - | 2 | 150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
