siRNA Details
Gene Symbol: TGIF2LX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138960.3 | NP_620410.3 | 2 | 771 | ||
GenBank mRNA | AF497480.1 | - | 1 | 751 | ||
BC117398.1 | - | 2 | 771 | 40125949 | ||
BC117400.1 | - | 2 | 771 | 40125951 |
Gene Symbol: TGIF2LY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139214.2 | NP_631960.1 | 2 | 785 | ||
GenBank mRNA | AF332223.1 | - | 2 | 785 | ||
AK292609.1 | - | 2 | 829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
