siRNA Details
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301870.1 | NP_001288799.1 | 6 | 898 | ||
NM_001301871.1 | NP_001288800.1 | 6 | 981 | |||
NM_019041.6 | NP_061914.3 | 6 | 1006 | |||
XM_017010959.1 | XP_016866448.1 | 7 | 5613 | |||
XM_017010960.1 | XP_016866449.1 | 7 | 1022 | |||
GenBank mRNA | AK027684.1 | - | 6 | 851 | ||
AK225970.1 | - | 6 | 880 | |||
AK290662.1 | - | 6 | 880 | |||
AK297671.1 | - | 6 | 873 | |||
BC011873.2 | - | 6 | 903 | 3029407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
