siRNA Details
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152344.3 | NP_689557.1 | 3 | 524 | ||
XM_011524309.2 | XP_011522611.1 | 2 | 288 | |||
GenBank mRNA | AK055218.1 | - | 2 | 252 | ||
AL832076.1 | - | 1 | 213 | |||
BC016791.2 | - | 1 | 25 | |||
BC044587.1 | - | 2 | 252 | 4794614 | ||
BC105288.1 | - | 2 | 266 | 6385345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
