siRNA Details
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152344.3 | NP_689557.1 | 5, 6 | 818 | ||
GenBank mRNA | AK055218.1 | - | 4, 5 | 546 | ||
AK300637.1 | - | 3, 4 | 258 | |||
AL832076.1 | - | 1 | 506 | |||
BC016791.2 | - | 1 | 319 | |||
BC044587.1 | - | 4, 5 | 546 | 4794614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
