siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 4, 5 | 430 | ||
XM_006716256.3 | XP_006716319.1 | 5, 6 | 525 | |||
XM_006716257.3 | XP_006716320.1 | 4, 5 | 453 | |||
XM_006716260.3 | XP_006716323.1 | 3, 4 | 339 | |||
XM_011543842.2 | XP_011542144.1 | 4 | 462 | |||
XM_011543843.2 | XP_011542145.1 | 4, 5 | 401 | |||
XM_017013916.1 | XP_016869405.1 | 4, 5 | 429 | |||
XM_017013917.1 | XP_016869406.1 | 3, 4 | 350 | |||
XM_017013918.1 | XP_016869407.1 | 3, 4 | 322 | |||
GenBank mRNA | BE729618.1 | - | 4, 5 | 372 |
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 3, 4 | 385 | ||
NM_201400.3 | NP_958802.1 | 5, 6 | 568 | |||
NM_201598.3 | NP_963892.1 | 4, 5 | 466 | |||
XM_005255157.4 | XP_005255214.1 | 4, 5 | 460 | |||
XM_005255158.4 | XP_005255215.1 | 6, 7 | 614 | |||
XM_011522404.2 | XP_011520706.1 | 5, 6 | 533 | |||
GenBank mRNA | AL522535.3 | - | 3, 4 | 307 | ||
AY037162.1 | - | 4, 5 | 434 | |||
BC010084.2 | - | 5, 6 | 488 | 2822323 | ||
BG024981.1 | - | 4, 5 | 433 | |||
BQ921410.1 | - | 5, 6 | 551 | |||
CA495371.1 | - | 4, 5 | 387 | |||
JX424572.1 | - | 2, 3 | 176 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 5, 6 | 487 | ||
XM_005273603.4 | XP_005273660.1 | 4, 5 | 460 | |||
XM_005273604.4 | XP_005273661.1 | 3, 4 | 379 | |||
XM_005273605.4 | XP_005273662.1 | 3, 4 | 280 | |||
XM_006716387.3 | XP_006716450.1 | 4, 5 | 481 | |||
XM_011544618.2 | XP_011542920.1 | 5, 6 | 532 | |||
XM_011544619.2 | XP_011542921.1 | 4, 5 | 499 | |||
XM_011544622.2 | XP_011542924.1 | 4, 5 | 401 | |||
XM_017013740.1 | XP_016869229.1 | 4 | 462 | |||
XM_017013741.1 | XP_016869230.1 | 3 | 399 | |||
XM_017013742.1 | XP_016869231.1 | 3 | 381 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 5, 6 | 591 | |||
GenBank mRNA | BC043342.2 | - | 4, 5 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
