siRNA Details
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 8 | 938 | ||
XM_005273603.4 | XP_005273660.1 | 7 | 911 | |||
XM_005273604.4 | XP_005273661.1 | 6 | 830 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 731 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 769 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 932 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 983 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 950 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 852 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 913 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 850 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 832 | |||
XM_017013743.1 | XP_016869232.1 | 5 | 532 | |||
GenBank mRNA | AK297741.1 | - | 5 | 513 | ||
AK298940.1 | - | 4 | 631 | |||
AK309984.1 | - | 7 | 725 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 881 | ||
NR_003494.1 | 6 | 615 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 976 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1046 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 790 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 633 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1055 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 994 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1022 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 943 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 915 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 504 | |||
GenBank mRNA | AK098138.1 | - | 6 | 582 | ||
AK308439.1 | - | 7 | 696 | |||
AK308513.1 | - | 5 | 741 | |||
AL529666.3 | - | 5 | 552 | |||
BC062615.1 | - | 6 | 659 | |||
BC085016.1 | - | 7 | 688 | |||
BC098427.1 | - | 6 | 665 | |||
DA552021.1 | - | 3 | 414 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 5 | 462 | ||
NM_018172.2 | NP_060642.2 | 5 | 483 | |||
NM_152563.2 | NP_689776.1 | 4 | 381 | |||
GenBank mRNA | AK001523.1 | - | 5 | 457 | ||
AK130709.1 | - | 4 | 378 | |||
AK293221.1 | - | 4 | 497 | |||
AK311428.1 | - | 5 | 439 | |||
AK311518.1 | - | 5 | 462 | |||
AK315454.1 | - | 4 | 355 | |||
BC032519.1 | - | 4 | 381 | 5494088 | ||
BQ921333.1 | - | 5 | 432 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 4 | 378 | |||
GenBank mRNA | AK055771.1 | - | 2 | 1031 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 8 | 1042 | |||
GenBank mRNA | BC043342.2 | - | 7 | 766 | ||
BC068108.1 | - | 6 | 575 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 4 | 1291 | |||
NR_130741.1 | 4 | 345 | ||||
NR_130742.1 | 4 | 295 | ||||
GenBank mRNA | BM718734.1 | - | 2 | 122 | ||
CN481473.1 | - | 3 | 206 | |||
HY065247.1 | - | 4 | 295 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 630 | |||
GenBank mRNA | BX282364.1 | - | 3 | 279 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 585 |
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 5 | 459 | |||
NR_024251.1 | 5 | 557 | ||||
GenBank mRNA | BC032772.1 | - | 5 | 445 | ||
BC041080.1 | - | 5 | 528 | |||
BM781991.1 | - | 5 | 459 |
Gene Symbol: FAM86KP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513917.2 | XP_011512219.1 | 4 | 355 | ||
XM_017008895.1 | XP_016864384.1 | 6 | 538 | |||
XM_017008896.1 | XP_016864385.1 | 3 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
