siRNA Details
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164356.1 | NP_001157828.1 | 2 | 127 | ||
NM_015963.5 | NP_057047.4 | 3 | 1570 | |||
XM_005247016.4 | XP_005247073.3 | 3 | 1803 | |||
XM_011511291.2 | XP_011509593.1 | 3 | 1372 | |||
XM_017004256.1 | XP_016859745.1 | 3 | 1364 | |||
GenBank mRNA | AF132970.1 | - | 2 | 127 | ||
AF258556.1 | - | 3 | 1322 | |||
AK001216.1 | - | 2 | 617 | |||
AK225608.1 | - | 2 | 95 | |||
BC000247.1 | - | 2 | 1012 | |||
BC000767.2 | - | 2 | 485 | |||
BC001842.2 | - | 2 | 485 | |||
BC009439.1 | - | 2 | 1108 | |||
BC069235.1 | - | 3 | 1500 | 6645198 | ||
BC071896.1 | - | 2 | 536 | |||
BC094822.1 | - | 2 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
