siRNA Details
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 19 | 2556 | ||
XM_017003011.1 | XP_016858500.1 | 19 | 2555 | |||
GenBank mRNA | AK294944.1 | - | 22 | 2439 | ||
DA203219.1 | - | 6 | 549 | |||
DA675097.1 | - | 6 | 524 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 2561 | |||
NM_001302371.1 | NP_001289300.1 | 2561 | ||||
GenBank mRNA | AF379632.1 | - | 2 | 87 | ||
AF379634.1 | - | 2 | 147 | |||
AF380580.1 | - | 2 | 83 | |||
AF419616.1 | - | 4 | 300 | |||
AL050141.1 | - | 2 | 1029 | |||
AY894565.1 | - | 18 | 2373 | |||
BX538005.1 | - | 27 | 3808 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 19 | 2526 | ||
XM_011509395.2 | XP_011507697.1 | 22 | 3026 | |||
GenBank mRNA | AK095459.1 | - | 5 | 1261 | ||
AL117237.1 | - | 13 | 1523 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 16 | 1986 | ||
GenBank mRNA | AF379633.1 | - | 4 | 386 | ||
AF379636.1 | - | 2 | 186 | |||
CR936830.1 | - | 4 | 3399 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 1881 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY894568.1 | - | 18 | 2329 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 20 | 2601 | ||
NM_001277444.1 | NP_001264373.1 | 24 | 3108 | |||
XM_006711317.2 | XP_006711380.1 | 25 | 3336 | |||
GenBank mRNA | AK294414.1 | - | 18 | 2266 | ||
AY894574.1 | - | 20 | 2603 | |||
BQ436001.1 | - | 4 | 335 | |||
DQ786323.1 | - | 1 | 42 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
