siRNA Details
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 7 | 953 | ||
NM_001277444.1 | NP_001264373.1 | 7 | 953 | |||
XM_006711317.2 | XP_006711380.1 | 8 | 1181 | |||
XM_017001278.1 | XP_016856767.1 | 8 | 1181 | |||
GenBank mRNA | AK294414.1 | - | 1 | 111 | ||
AY894574.1 | - | 7 | 955 | |||
BC071995.1 | - | 2 | 137 | |||
BC094841.1 | - | 2 | 137 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 913 | |||
NM_001302371.1 | NP_001289300.1 | 913 | ||||
GenBank mRNA | BX538005.1 | - | 12 | 2048 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 6 | 878 | ||
XM_011509395.2 | XP_011507697.1 | 9 | 1378 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 8 | 1227 | ||
NM_173638.4 | NP_775909.2 | 5 | 697 | |||
XM_017001077.1 | XP_016856566.1 | 3 | 345 | |||
GenBank mRNA | AK097180.1 | - | 8 | 1146 | ||
AK290302.1 | - | 2 | 156 | |||
AY894566.1 | - | 2 | 137 | |||
BC023087.1 | - | 5 | 554 | 3918932 | ||
CV024999.1 | - | 1 | 65 | |||
ES313907.1 | - | 2 | 256 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 6 | 908 | ||
XM_017003011.1 | XP_016858500.1 | 6 | 907 |
Gene Symbol: NBPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047980.2 | NP_001041445.1 | 1 | 470 | ||
XM_017001186.1 | XP_016856675.1 | 1 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
