siRNA Details
Gene Symbol: KLHL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031710.2 | NP_001026880.2 | 4 | 633 | ||
NM_001172428.1 | NP_001165899.1 | 4 | 633 | |||
NM_018846.4 | NP_061334.4 | 4 | 717 | |||
NR_033328.1 | 5 | 764 | ||||
NR_033329.1 | 5 | 764 | ||||
XM_006715753.2 | XP_006715816.1 | 4 | 628 | |||
XM_006715754.2 | XP_006715817.1 | 5 | 711 | |||
XM_006715755.2 | XP_006715818.1 | 5 | 930 | |||
XM_006715756.2 | XP_006715819.1 | 4 | 799 | |||
XM_006715757.3 | XP_006715820.1 | 4 | 628 | |||
XM_017012439.1 | XP_016867928.1 | 5 | 930 | |||
XM_017012440.1 | XP_016867929.1 | 4 | 628 | |||
XM_017012441.1 | XP_016867930.1 | 5 | 711 | |||
GenBank mRNA | AF111113.1 | - | 4 | 517 | ||
AK056390.1 | - | 5 | 715 | |||
AK290479.1 | - | 4 | 427 | |||
AK297595.1 | - | 3 | 474 | |||
AK299006.1 | - | 4 | 359 | |||
AK302995.1 | - | 5 | 487 | |||
AL136597.1 | - | 4 | 579 | |||
AL834193.1 | - | 1 | 375 | |||
AY028802.1 | - | 4 | 282 | |||
BC009555.1 | - | 5 | 680 | |||
BC039585.1 | - | 4 | 575 | 5527346 | ||
BG704835.1 | - | 4 | 724 | |||
EF560731.1 | - | 5 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
