siRNA Details
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 4 | 490 | ||
NR_033249.1 | 3 | 426 | ||||
XM_017023136.1 | XP_016878625.1 | 4 | 489 | |||
XM_017023137.1 | XP_016878626.1 | 4 | 432 | |||
GenBank mRNA | AK311978.1 | - | 4 | 399 | ||
AY533031.1 | - | 1 | 249 | |||
BC000790.1 | - | 1 | 414 | 3451361 | ||
BC009065.1 | - | 1 | 449 | |||
BC020922.1 | - | 1 | 453 | 4247157 | ||
BF798008.1 | - | 4 | 361 | |||
BG708388.1 | - | 4 | 523 | |||
BT009827.1 | - | 4 | 393 | |||
CN283805.1 | - | 3 | 316 | |||
CN283809.1 | - | 4 | 523 | |||
CR457072.1 | - | 4 | 393 | |||
D00723.1 | - | 1 | 485 | |||
M69175.1 | - | 4 | 417 |
Gene Symbol: GCSHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033248.1 | 1 | 334 | |||
GenBank mRNA | AK092810.1 | - | 2 | 2218 | ||
AV703937.1 | - | 1 | 463 | |||
BM685716.1 | - | 1 | 87 |
Gene Symbol: LOC729080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033244.1 | 1 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
