siRNA Details
Gene Symbol: GCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004483.4 | NP_004474.2 | 5 | 588 | ||
NR_033249.1 | 4 | 524 | ||||
XM_017023136.1 | XP_016878625.1 | 5 | 605 | |||
XM_017023137.1 | XP_016878626.1 | 5 | 548 | |||
GenBank mRNA | AK311978.1 | - | 5 | 497 | ||
AY533031.1 | - | 1 | 347 | |||
BC000790.1 | - | 1 | 512 | 3451361 | ||
BC009065.1 | - | 1 | 547 | |||
BC020922.1 | - | 1 | 551 | 4247157 | ||
BF798008.1 | - | 5 | 459 | |||
BG708388.1 | - | 5 | 621 | |||
BT009827.1 | - | 5 | 491 | |||
CN283805.1 | - | 4 | 412 | |||
CN283809.1 | - | 5 | 621 | |||
CR457072.1 | - | 5 | 491 | |||
D00723.1 | - | 1 | 583 | |||
M69175.1 | - | 5 | 515 |
Gene Symbol: GCSHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033248.1 | 1 | 432 | |||
GenBank mRNA | AK092810.1 | - | 2 | 2316 | ||
AV703937.1 | - | 1 | 561 | |||
BM685716.1 | - | 1 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
